Ethyl-3-cyclopentene-1-carboxylate ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
E0831-25G E0831-5G
TCIAE0831-25GEA 236 EUR
TCIAE0831-25G TCIAE0831-5G
Ethyl-3-cyclopentene-1-carboxylate ≥97.0% (by GC)
Ethyl-3-cyclopentene-1-carboxylate
Formule: C₈H₁₂O₂
Kookpunt: 64°C
MDL Nummer: MFCD06410819
CAS nummer: 21622-01-5
UN: 3272
ADR: 3,III
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %