Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Ethyl 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylate, 2,4-Dihydroxy-6-methyl-3-pyridinecarboxylic acid ethyl ester, Ethyl 2,4-dihydroxy-6-methyl-3-pyridinecarboxylate, Ethyl 2,4-Dihydroxy-6-methylnicotinate, 2,4-Dihydroxy-6-methylnicotinic acid ethyl ester

E0869-1G E0869-5G
TCIAE0869-1GEA 104 EUR
TCIAE0869-1G TCIAE0869-5G
Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate ≥98.0% (by GC, titration analysis)
Ethyl-2,4-dihydroxy-6-methyl-3-pyridinecarboxylate
Formule: C₉H₁₁NO₄
Smeltpunt: 210°C
MDL Nummer: MFCD02929397
CAS nummer: 70254-52-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR