Ethyl pyrimidine-4-carboxylate ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Ethyl 4-pyrimidinecarboxylate

E0881-5G E0881-25G
TCIAE0881-5GEA 131 EUR
TCIAE0881-5G TCIAE0881-25G
Ethyl pyrimidine-4-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl pyrimidine-4-carboxylate
Formule: C₇H₈N₂O₂
Kookpunt: 160°C
MDL Nummer: MFCD09832999
CAS nummer: 62846-82-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR