Estra-4,9-diene-3,17-dione ≥95.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E0920-5G E0920-1G
TCIAE0920-5GEA 200 EUR
TCIAE0920-5G TCIAE0920-1G
Estra-4,9-diene-3,17-dione ≥95.0% (by HPLC)
Estra-4,9-diene-3,17-dione
Formule: C₁₈H₂₂O₂
MW: 270.37 g/mol
MDL Nummer: MFCD03695567
CAS nummer: 5173-46-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 137.0 to 144.0 deg-C
Specific rotation [a]20/D -190.0 to -200.0 deg (C=0.5、MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR