Ethyl-2,4-difluorobenzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E0935-5G E0935-25G
TCIAE0935-5GEA 17.6 EUR
TCIAE0935-5G TCIAE0935-25G
Ethyl-2,4-difluorobenzoate ≥98.0% (by GC)
Ethyl-2,4-difluorobenzoate
Formule: C₉H₈F₂O₂
Kookpunt: 88°C
MDL Nummer: MFCD00153149
CAS nummer: 108928-00-3

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR