(±)-Ethyl-2-bromovalerate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Ethyl 2-bromovalerate
E0970-25G
E0970-100G
TCIAE0970-25GEA
35.2
EUR
TCIAE0970-25G
TCIAE0970-100G
(±)-Ethyl-2-bromovalerate ≥98.0% (by GC)
(±)-Ethyl-2-bromovalerate
Formule:
C₇H₁₃BrO₂ MW: 209.09 g/mol Kookpunt: 94°C Vlampunt: 77°C |
MDL Nummer:
MFCD00000159 CAS nummer: 615-83-8 EINECS: 210-450-1 UN: 3265 ADR: 8 III |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...