1-Ethyl-1-methylpyrrolidiniumtetrafluoroborate ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

E0977-5G E0977-25G
TCIAE0977-5GEA 61.6 EUR
TCIAE0977-5G TCIAE0977-25G
1-Ethyl-1-methylpyrrolidiniumtetrafluoroborate ≥98.0% (by total nitrogen basis)
1-Ethyl-1-methylpyrrolidiniumtetrafluoroborate
Formule: C₇H₁₆BF₄N
MDL Nummer: MFCD08458505
CAS nummer: 117947-85-0
UN: 1759
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR