Ethyl-4-phenylbutyrate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1027-5G
E1027-1G
TCIAE1027-5GEA
74.8
EUR
TCIAE1027-5G
TCIAE1027-1G
Ethyl-4-phenylbutyrate ≥98.0% (by GC)
Ethyl-4-phenylbutyrate
Formule:
C₁₂H₁₆O₂ Kookpunt: 100°C |
MDL Nummer:
MFCD00026923 CAS nummer: 10031-93-3 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...