Ethyl 2-Chloro-6-fluorophenylacetate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1038-1G
E1038-5G
TCIAE1038-1GEA
90.2
EUR
TCIAE1038-1G
TCIAE1038-5G
Ethyl 2-Chloro-6-fluorophenylacetate ≥98.0% (by GC)
Ethyl 2-Chloro-6-fluorophenylacetate
Formule:
C₁₀H₁₀ClFO₂ |
MDL Nummer:
MFCD00800606 CAS nummer: 214262-85-8 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...