Ethylamine hydroiodide ≥98.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

E1045-1G E1045-5G
TCIAE1045-1GEA 57.2 EUR
TCIAE1045-1G TCIAE1045-5G
Ethylamine hydroiodide ≥98.0% (by total nitrogen and titration analysis)
Ethylamine hydroiodide
Formule: C₂H₇N
CAS nummer: 506-58-1
Merck Index: 14,03762

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 189.0 to 193.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR