Ethylene Glycol 1,2-Bis(2-propynyl) ether ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Bis-propargyl-PEG2
E1054-5G E1054-25G
TCIAE1054-5GEA 135 EUR
TCIAE1054-5G TCIAE1054-25G
Ethylene Glycol 1,2-Bis(2-propynyl) ether ≥97.0% (by GC)
Ethylene Glycol 1,2-Bis(2-propynyl) ether
Formule: C₈H₁₀O₂
MW: 138.17 g/mol
Kookpunt: 88°C
MDL Nummer: MFCD03640302
CAS nummer: 40842-04-4
UN: 3271
ADR: 3 III
Order Now
Specification Test Results
Appearance White to Yellow to Green clear liquid
Purity (GC) min. 97.0 %