Ethyl 2-Oxocycloheptanecarboxylate ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E1056-1G E1056-200MG
TCIAE1056-1GEA 251 EUR
TCIAE1056-1G TCIAE1056-200MG
Ethyl 2-Oxocycloheptanecarboxylate ≥95.0% (by GC)
Ethyl 2-Oxocycloheptanecarboxylate
Formule: C₁₀H₁₆O₃
Kookpunt: 126°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00604086
CAS nummer: 774-05-0

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR