Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

E1087-5G E1087-25G
TCIAE1087-5GEA 37.4 EUR
TCIAE1087-5G TCIAE1087-25G
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate ≥96.0% (by GC)
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate
Formule: C₉H₁₁F₃O₄
Kookpunt: 120°C
Vlampunt: 104°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD02677683
CAS nummer: 571-55-1

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR