Ethyl-5-chlorothiophene-2-carboxylate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1112-1G E1112-5G
TCIAE1112-1GEA 28.6 EUR
TCIAE1112-1G TCIAE1112-5G
Ethyl-5-chlorothiophene-2-carboxylate ≥98.0% (by GC)
Ethyl-5-chlorothiophene-2-carboxylate
Formule: C₇H₇ClO₂S
Kookpunt: 62°C
MDL Nummer: MFCD00051732
CAS nummer: 5751-82-6
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %