Ethyl-4-acetylbenzoate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E1113-1G E1113-5G
TCIAE1113-1GEA 39.6 EUR
TCIAE1113-1G TCIAE1113-5G
Ethyl-4-acetylbenzoate ≥97.0% (by GC)
Ethyl-4-acetylbenzoate
Formule: C₁₁H₁₂O₃
MDL Nummer: MFCD00013241
CAS nummer: 38430-55-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR