Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1123-1G
E1123-5G
TCIAE1123-1GEA
57.2
EUR
TCIAE1123-1G
TCIAE1123-5G
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate ≥98.0% (by HPLC, titration analysis)
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate
Formule:
C₁₁H₁₀N₂O₃ |
MDL Nummer:
MFCD00051583 CAS nummer: 29113-33-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 190.0 to 194.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...