Ethyl 2-Iodopropionate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

E1198-5G E1198-25G
TCIAE1198-5GEA 49.5 EUR
TCIAE1198-5G TCIAE1198-25G
Ethyl 2-Iodopropionate ≥97.0% (by GC)
Ethyl 2-Iodopropionate
Formule: C₅H₉IO₂
Kookpunt: 79°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD02258628
CAS nummer: 31253-08-4

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR