Ethyl-3,5-dimethylisoxazole-4-carboxylate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E1212-1G E1212-5G
TCIAE1212-1GEA 20.9 EUR
TCIAE1212-1G TCIAE1212-5G
Ethyl-3,5-dimethylisoxazole-4-carboxylate ≥98.0% (by GC)
Ethyl-3,5-dimethylisoxazole-4-carboxylate
Formule: C₈H₁₁NO₃
MW: 169.18 g/mol
Kookpunt: 220°C
MDL Nummer: MFCD00052557
CAS nummer: 17147-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da2e513392b9add12e838d2055f89a90757c8824&vid=da2e513392b9add12e838d2055f89a90757c8824&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR