Ethyl-6-aminopicolinate ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
6-Aminopyridine-2-carboxylic acid ethyl ester, Ethyl 6-Aminopicolinate, Ethyl 6-Aminopyridine-2-carboxylate, 6-Aminopicolinic acid ethyl ester, Ethyl 6-aminopicolinate
E1213-1G
E1213-5G
TCIAE1213-1GEA
62.7
EUR
TCIAE1213-1G
TCIAE1213-5G
Ethyl-6-aminopicolinate ≥98.0% (by GC, titration analysis)
Ethyl-6-aminopicolinate
Formule:
C₈H₁₀N₂O₂ MW: 166.1800 g/mol |
MDL Nummer:
MFCD01009842 CAS nummer: 69142-64-9 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 70.0 to 74.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...