Ethylenediamine dihydrobromide ≥97.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

E1221-1G E1221-5G
TCIAE1221-1GEA 61.6 EUR
TCIAE1221-1G TCIAE1221-5G
Ethylenediamine dihydrobromide ≥97.0% (by total nitrogen and titration analysis)
Ethylenediamine dihydrobromide
Formule: C₂H₈N₂·₂HBr
Smeltpunt: 350°C
CAS nummer: 624-59-9
EINECS: 210-853-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR