1-Ethyl-3,5-difluorobenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
E1229-5G
E1229-25G
TCIAE1229-5GEA
38.5
EUR
TCIAE1229-5G
TCIAE1229-25G
1-Ethyl-3,5-difluorobenzene ≥98.0% (by GC)
1-Ethyl-3,5-difluorobenzene
Formule:
C₈H₈F₂ Vlampunt: 31°C |
MDL Nummer:
MFCD10000628 CAS nummer: 117358-52-8 UN: 1993 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...