Ethyl-1-bromocyclobutanecarboxylate ≥96.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
E1245-1G
E1245-5G
TCIAE1245-1GEA
34.1
EUR
TCIAE1245-1G
TCIAE1245-5G
Ethyl-1-bromocyclobutanecarboxylate ≥96.0% (by GC)
Ethyl-1-bromocyclobutanecarboxylate
Formule:
C₇H₁₁BrO₂ Kookpunt: 88°C Vlampunt: 79°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00001322 CAS nummer: 35120-18-4 EINECS: 252-384-6 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...