Ethyl 2-iodobenzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Ethyl 2-iodobenzoate, 2-Iodobenzoic acid ethyl ester

E1273-5G E1273-25G
TCIAE1273-5GEA 27.5 EUR
TCIAE1273-5G TCIAE1273-25G
Ethyl 2-iodobenzoate ≥98.0% (by GC)
Ethyl-2-iodobenzoate
Formule: C₉H₉IO₂
Kookpunt: 165°C
MDL Nummer: MFCD00051796
CAS nummer: 1829-28-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36d58e235a70a0a1412b5cbb00b1b58f03c718b9&vid=36d58e235a70a0a1412b5cbb00b1b58f03c718b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR