1-Ethyl-3-methylimidazolium tricyanomethanide ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

E1298-5G E1298-25G
TCIAE1298-5GEA 79.2 EUR
TCIAE1298-5G TCIAE1298-25G
1-Ethyl-3-methylimidazolium tricyanomethanide ≥98.0% (by HPLC, total nitrogen)
1-Ethyl-3-methylimidazolium tricyanomethanide
Formule: C₁₀H₁₁N₅
Smeltpunt: -10°C
MDL Nummer: MFCD22460460
CAS nummer: 666823-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44b43a0c568efa696fdfdec306466ce131257349&vid=44b43a0c568efa696fdfdec306466ce131257349&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR