Ethyl 2-(4-Cyanophenyl)hydrazinecarboxylate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
E1299-1G
E1299-5G
TCIAE1299-1GEA
58.3
EUR
TCIAE1299-1G
TCIAE1299-5G
Ethyl 2-(4-Cyanophenyl)hydrazinecarboxylate ≥98.0% (by GC)
Ethyl 2-(4-Cyanophenyl)hydrazinecarboxylate
Formule:
C₁₀H₁₁N₃O₂ |
CAS nummer:
700371-70-6 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...