1-Ethyl-3-methylimidazolium diethylphosphate ≥96.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1-ethyl-3-methylimidazolium diethylphosphate
E1309-5G
E1309-25G
TCIAE1309-5GEA
28.6
EUR
TCIAE1309-5G
TCIAE1309-25G
1-Ethyl-3-methylimidazolium diethylphosphate ≥96.0% (by HPLC, titration analysis)
1-Ethyl-3-methylimidazolium diethylphosphate
Formule:
C₁₀H₂₁N₂O₄P |
MDL Nummer:
MFCD09953486 CAS nummer: 848641-69-0 UN: 3265 ADR: 8,II |
|
Specification Test Results
Appearance | Colorless to Yellow to Orange clear liquid |
Purity (HPLC) | min. 96.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...