1-Ethynyl-3,5-bis(trifluoromethyl)benzene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
3',5'-Bis(trifluoromethyl)phenylacetylene
E1353-1G
E1353-5G
TCIAE1353-1GEA
45.1
EUR
TCIAE1353-1G
TCIAE1353-5G
1-Ethynyl-3,5-bis(trifluoromethyl)benzene ≥98.0% (by GC)
1-Ethynyl-3,5-bis(trifluoromethyl)benzene
Formule:
C₁₀H₄F₆ Kookpunt: 74°C Vlampunt: 44°C |
MDL Nummer:
MFCD01861850 CAS nummer: 88444-81-9 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...