2-Ethynyl-1,3,5-trimethylbenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
1-Ethynyl-2,4,6-trimethylbenzene, 2-Ethynylmesitylene, 2,4,6-Trimethylphenylacetylene
E1361-1G
E1361-5G
TCIAE1361-1GEA
66
EUR
TCIAE1361-1G
TCIAE1361-5G
2-Ethynyl-1,3,5-trimethylbenzene ≥98.0% (by GC)
2-Ethynyl-1,3,5-trimethylbenzene
Formule:
C₁₁H₁₂ Kookpunt: 215°C Smeltpunt: 4°C Vlampunt: 83°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD07777099 CAS nummer: 769-26-6 |
Specification Test Results
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...