Ectoine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI EUROPE
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TCIAE1372-1GEA 68.2 EUR
TCIAE1372-1G
Ectoine ≥98.0% (by HPLC, titration analysis)
Ectoine
Formule: C₆H₁₀N₂O₂
MW: 142,1558 g/mol
MDL Nummer: MFCD03419286
CAS nummer: 96702-03-3
Merck Index: 14,03505

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +165 to +175 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR