Ethyl 2-(4-Oxocyclohexyl)acetate ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

E1375-1G E1375-5G
TCIAE1375-1GEA 39.6 EUR
TCIAE1375-1G TCIAE1375-5G
Ethyl 2-(4-Oxocyclohexyl)acetate ≥95.0% (by GC)
Ethyl 2-(4-Oxocyclohexyl)acetate
Formule: C₁₀H₁₆O₃
Kookpunt: 96°C
MDL Nummer: MFCD11045783
CAS nummer: 58012-34-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR