1-Ethyl-2-methylindole ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1376-25G E1376-5G
TCIAE1376-25GEA 262 EUR
TCIAE1376-25G TCIAE1376-5G
1-Ethyl-2-methylindole ≥98.0% (by GC)
1-Ethyl-2-methylindole
Formule: C₁₁H₁₃N
Kookpunt: 270°C
MDL Nummer: MFCD01632168
CAS nummer: 40876-94-6
EINECS: 255-121-3
Order Now
Specification Test Results
Appearance White to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %