1-Ethyl-4-isopropylbenzene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1406-1ML
E1406-5ML
TCIAE1406-1MLEA
90.2
EUR
TCIAE1406-1ML
TCIAE1406-5ML
1-Ethyl-4-isopropylbenzene ≥98.0% (by GC)
1-Ethyl-4-isopropylbenzene
Formule:
C₁₁H₁₆ Smeltpunt: 197°C |
MDL Nummer:
MFCD00061006 CAS nummer: 4218-48-8 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...