Ferroin (1,10-phenanthrolineferrous sulphate complex) solution_SC acc. to JIS K8001
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate
F0005-25G F0005-500G
TCIAF0005-25GEA 17.6 EUR
TCIAF0005-25G TCIAF0005-500G
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution_SC acc. to JIS K8001
Ferroin (1,10-phenanthrolineferrous sulphate complex)
Formule: C₃₆H₂₄FeN₆SO₄
MW: 692.54 g/mol
Dichtheid: 0.9999
MDL Nummer: MFCD00036428
CAS nummer: 14634-91-4
EINECS: 238-676-6
Order Now
Specification Test Results
Appearance Orange to Brown to Dark red clear liquid to cloudy liquid