2-Furoyltrifluoroacetone

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

F0083-25G F0083-5G
TCIAF0083-25GEA 97.9 EUR
TCIAF0083-25G TCIAF0083-5G
2-Furoyltrifluoroacetone
2-Furoyltrifluoroacetone
Formule: C₈H₅F₃O₃
Kookpunt: 84°C
MDL Nummer: MFCD00020935
CAS nummer: 326-90-9
EINECS: 206-315-1

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Freezing point 20.0 to 22.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR