(Ferrocenylmethyl)trimethylammonium bromide ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

F0166-1G F0166-5G
TCIAF0166-1GEA 257 EUR
TCIAF0166-1G TCIAF0166-5G
(Ferrocenylmethyl)trimethylammonium bromide ≥95.0% (by titrimetric analysis)
(Ferrocenylmethyl)trimethylammonium bromide
Formule: C₁₄H₂₀BrFeN
MDL Nummer: MFCD00059981
CAS nummer: 106157-30-6

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (Chelometric Titration) min. 95.0 %
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR