Methyl fumaraldehydate ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Fumaraldehydic Acid Methyl Ester, Fumaraldehydicacidmethylester, 3-(Methoxycarbonyl)-2-propenal, 3-(Methoxycarbonyl)acrylaldehyde

F0351-1G F0351-100MG
TCIAF0351-1GEA 546 EUR
TCIAF0351-1G TCIAF0351-100MG
Methyl fumaraldehydate ≥96.0% (by GC)
Methyl fumaraldehydate
Formule: C₅H₆O₃
MW: 114.10 g/mol
Kookpunt: 88°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00191651
CAS nummer: 5837-72-9

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR