Ferroceneacetic acid ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

F0406-1G F0406-5G
TCIAF0406-1GEA 196 EUR
TCIAF0406-1G TCIAF0406-5G
Ferroceneacetic acid ≥97.0% (by GC)
Ferroceneacetic acid
Formule: C₁₂H₁₂FeO₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00045464
CAS nummer: 1287-16-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2968a185856e0df5ab820bbcbea749e27f050c2a&vid=2968a185856e0df5ab820bbcbea749e27f050c2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 97.0 %
Purity (Chelometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR