N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine hydrate ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

F0454-1G F0454-5G
TCIAF0454-1GEA 17.6 EUR
TCIAF0454-1G TCIAF0454-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine hydrate ≥97.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine hydrate
Formule: C₁₈H₁₇NO₅
MDL Nummer: MFCD04113634
CAS nummer: 73724-45-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b1ac80311e84df1d89bbba3fae0703cc6457452&vid=e5898ec04ba2cbe50228119c8a3c20ad92e7c2a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 % (calcd.on anh.substance)
Specific rotation [a]20/D -10.0 to -15.0 deg (C=1, DMF)
Water max. 6.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR