Formylhydrazine ≥97.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Formic acid hydrazide, Formohydrazide, Formic hydrazide

F0667-25G F0667-100G
TCIAF0667-25GEA 61.6 EUR
TCIAF0667-25G TCIAF0667-100G
Formylhydrazine ≥97.0% (by HPLC)
Formylhydrazine
Formule: CH₄N₂O
MW: 60.06 g/mol
Kookpunt: 90°C
Smeltpunt: 150
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007589
CAS nummer: 624-84-0
EINECS: 210-867-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a627802d86fc30ee6d8aeeafe8375946aab3f0e6&vid=a627802d86fc30ee6d8aeeafe8375946aab3f0e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 56.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR