9-Fluorenylmethyl carbamate ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Carbamic Acid 9-Fluorenylmethyl Ester, Fmoc-NH2, (9H-Fluoren-9-ylmethoxy)carboxamide

F0689-1G F0689-5G
TCIAF0689-1GEA 56.1 EUR
TCIAF0689-1G TCIAF0689-5G
9-Fluorenylmethyl carbamate ≥98.0% (by HPLC, total nitrogen)
9-Fluorenylmethyl carbamate
Formule: C₁₅H₁₃NO₂
MDL Nummer: MFCD00237376
CAS nummer: 84418-43-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR