Fexofenadine hydrochloride ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F0698-1G
F0698-5G
TCIAF0698-1GEA
101
EUR
TCIAF0698-1G
TCIAF0698-5G
Fexofenadine hydrochloride ≥98.0% (by HPLC, titration analysis)
Fexofenadine hydrochloride
Formule:
C₃₂H₃₉NO₄ Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD00865710 CAS nummer: 153439-40-8 Merck Index: 14,04068 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...