3-(4-Fluorophenyl)-1-isopropylindole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 3-(4-Fluorophenyl)-1-isopropyl-1H-indole

F0743-25G F0743-5G
TCIAF0743-25GEA 180 EUR
TCIAF0743-25G TCIAF0743-5G
3-(4-Fluorophenyl)-1-isopropylindole ≥98.0% (by GC)
3-(4-Fluorophenyl)-1-isopropylindole
Formule: C₁₇H₁₆FN
MW: 253.32 g/mol
MDL Nummer: MFCD01075733
CAS nummer: 93957-49-4
EINECS: 418-790-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR