Fluvastatin sodium salt ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: XU 62-320, rel-(3S,5R,E)-7-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoic acid sodium salt, Sodium rel-(3R,5S,E)-7-(3-(4-fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoate

F0820-5G F0820-1G
TCIAF0820-5GEA 257 EUR
TCIAF0820-5G TCIAF0820-1G
Fluvastatin sodium salt ≥98.0% (by HPLC, total nitrogen)
Fluvastatin sodium salt
Formule: C₂₄H₂₅FNNaO₄
MW: 433.46 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00929076
CAS nummer: 93957-55-2
Merck Index: 14,04218

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Water max. 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR