N-Formylsaccharin ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F0854-5G
F0854-25G
TCIAF0854-5GEA
77
EUR
TCIAF0854-5G
TCIAF0854-25G
N-Formylsaccharin ≥98.0% (by GC)
N-Formylsaccharin
Formule:
C₈H₅NO₄S |
MDL Nummer:
MFCD23380208 CAS nummer: 50978-45-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...