Fluvoxamine maleate ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone O-(2-Aminoethyl)oxime Maleate

F0858-1G F0858-5G
TCIAF0858-1GEA 64.9 EUR
TCIAF0858-1G TCIAF0858-5G
Fluvoxamine maleate ≥98.0% (by HPLC, titration analysis)
Fluvoxamine maleate
Formule: C₁₅H₂₁F₃N₂O₂
MW: 434.41 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00269809
CAS nummer: 61718-82-9
Merck Index: 14,04219

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR