Formononetin ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

F0868-1G F0868-5G
TCIAF0868-1GEA 78.1 EUR
TCIAF0868-1G TCIAF0868-5G
Formononetin ≥98.0% (by HPLC, titration analysis)
Formononetin
Formule: C₁₆H₁₂O₄
MDL Nummer: MFCD00016948
CAS nummer: 485-72-3
EINECS: 207-623-9
Merck Index: 14,04244

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 256.0 to 260.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR