Nα-Fmoc-Nω-(p-toluenesulphonyl)-L-arginine ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

F0883-5G F0883-1G
TCIAF0883-5GEA 84.7 EUR
TCIAF0883-5G TCIAF0883-1G
Nα-Fmoc-Nω-(p-toluenesulphonyl)-L-arginine ≥98.0% (by HPLC)
Nα-Fmoc-Nω-(p-toluenesulphonyl)-L-arginine
Formule: C₂₈H₃₀N₄O₆S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00063331
CAS nummer: 83792-47-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %
Melting point 92.0 to 96.0 deg-C
Specific rotation [a]20/D -5.0 to -7.0 deg (C=1、DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR