4-Fluoro-3-nitrophenyl methyl sulphone ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Fluoro-5-methylsulphonylnitrobenzene, 1-Fluoro-4-(methylsulphonyl)-2-nitrobenzene

F0887-1G F0887-5G
TCIAF0887-1GEA 66 EUR
TCIAF0887-1G TCIAF0887-5G
4-Fluoro-3-nitrophenyl methyl sulphone ≥98.0% (by GC)
4-Fluoro-3-nitrophenyl methyl sulphone
Formule: C₇H₆FNO₄S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00025066
CAS nummer: 453-72-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 156.0 to 160.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR