5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

F0939-1G F0939-5G
TCIAF0939-1GEA 33 EUR
TCIAF0939-1G TCIAF0939-5G
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
Formule: C8H9NO3
Smeltpunt: 283°C
MDL Nummer: MFCD06202342
CAS nummer: 253870-02-9

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR