2-Fluoro-m-toluic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Fluoro-3-methylbenzoic acid

F0949-5G F0949-1G
TCIAF0949-5GEA 149 EUR
TCIAF0949-5G TCIAF0949-1G
2-Fluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
2-Fluoro-m-toluic acid
Formule: C₈H₇FO₂
MDL Nummer: MFCD00042504
CAS nummer: 315-31-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR